Learn More
Bio Rad Laboratories CRIT XT BIS-TRIS GEL 4-12% 12W
Catalog No. NC9070127
Encompass
Criterion XT Bis-Tris Gel, 12 + 2 Well, 45microliter, Dimensions: 13.3W x 8.7cm long, Resolving Gel (%T): 4 to 12%, Crosslinker (%C): 5%, XT MES buffer: 2.5 to 200 kd, XT MOPS buffer: 10 to 300 kd
Catalog No. NC9070127 Supplier Bio Rad Laboratories Supplier No. 3450123
Criterion XT Bis-Tris Gel, 12 + 2 Well, 45microliter, Dimensions: 13.3W x 8.7cm long, Resolving Gel (%T): 4 to 12%, Crosslinker (%C): 5%, XT MES buffer: 2.5 to 200 kd, XT MOPS buffer: 10 to 300 kd