Learn More
Bio Rad Laboratories CRIT XT BIS-TRIS GEL 4-12% 30U
Catalog No. NC9038759
Encompass
Criterion XT Bis-Tris Gel, 4-12%, 18-Well, 30 microliter, 13.3W x 8.7 cm long
Catalog No. NC9038759 Supplier Bio Rad Laboratories Supplier No. 3450124
Criterion XT Bis-Tris Gel, 4-12%, 18-Well, 30 microliter, 13.3W x 8.7 cm long