Learn More
Bio Rad Laboratories 4–15% Criterion Tris-HCl Protein Gel, 18 well, 30 uL
Catalog No. NC1642033
Encompass
Pkg of 1, 4–15% precast polyacrylamide gel, 13.3 × 8.7 cm (W × L), for use with Criterion and Criterion Dodeca Electrophoresis Cells.
Catalog No. NC1642033 Supplier Bio Rad Laboratories Supplier No. 3450028
Pkg of 1, 4–15% precast polyacrylamide gel, 13.3 × 8.7 cm (W × L), for use with Criterion and Criterion Dodeca Electrophoresis Cells.