Learn More
Bio Rad Laboratories CRITERION EMPTY CASSETTES
Supplier:  Bio Rad Laboratories 3459901
Encompass
Cassettes, Empty; Bio-Rad; Criterion; Accessory; 1.0mm thick with 12+2 well comb; 10 sets
Catalog No. 50-901-2283