Learn More
Bio Rad Laboratories CRITERION GEL 4-15
Catalog No. NC9015237
Encompass
Criterion Tris-HCl Gel, 4 to 15%, 12+2 Well, 45 microliter, 13.3W x 8.7 cm long
Catalog No. NC9015237 Supplier Bio Rad Laboratories Supplier No. 3450027
Criterion Tris-HCl Gel, 4 to 15%, 12+2 Well, 45 microliter, 13.3W x 8.7 cm long