Learn More
Bio Rad Laboratories CRITERION TGX PRECAST GELS 4-
Catalog No. 507513663
Encompass
Criterion TGX are long shelf-life Laemmli-like precast polyacrylamide gels for vertical SDS-PAGE and Native PAGE protein separations. Gel size is 13.3 x 8.7 x 0.1 cm (W x L x Thickness) and are supplied in a 15.0 x 10.6 x 0.53 cm.
Catalog No. 50-751-3663 Supplier Bio Rad Laboratories Supplier No. 5671084
Criterion TGX are long shelf-life Laemmli-like precast polyacrylamide gels for vertical SDS-PAGE and Native PAGE protein separations. Gel size is 13.3 x 8.7 x 0.1 cm (W x L x Thickness) and are supplied in a 15.0 x 10.6 x 0.53 cm.