Learn More
Bio Rad Laboratories CRITERION XT BIS-TRIS GEL
Catalog No. 509012794
Encompass
Gel, XT Bis-Tris; Bio-Rad; 12 months shelf life; 4-12%; Prep+2 well; 800uL; W x L: 13.3 x 8.7cm
Catalog No. 50-901-2794 Supplier Bio Rad Laboratories Supplier No. 3450126
Gel, XT Bis-Tris; Bio-Rad; 12 months shelf life; 4-12%; Prep+2 well; 800uL; W x L: 13.3 x 8.7cm