Learn More
Bio Rad Laboratories CRITERION XT TRIS-ACETATE GEL
Catalog No. 509012847
Encompass
Gel, XT Tris-Acetate; Bio-Rad; 8-month shelf life; 3-8%; 12+2 well; 45uL; W x L: 13.3 x 8.7cm
Catalog No. 50-901-2847 Supplier Bio Rad Laboratories Supplier No. 3450129
Gel, XT Tris-Acetate; Bio-Rad; 8-month shelf life; 3-8%; 12+2 well; 45uL; W x L: 13.3 x 8.7cm