Learn More
AlphaProTech Critical Cover™ CoolOne™ Sensitive Skin Face Masks
BIOPHARMA
Catalog No. 19166634 Shop All AlphaProTech Products
Change view
Click to view available options
Includes:
Anti-fog earloops
Soft knit earloops
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-166-634 Anti-fog earloops
19-140-223 Soft knit earloops
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-166-634 Supplier AlphaProTech Supplier No. WH 6155
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Fluid resistant face masks are free of pigments and dyes and are available in two earloop styles. Patented Magic Arch™ support strip holds the mask away from the nose and mouth to create a larger breathing chamber.
  • Fluid resistant face masks are free of pigments and dyes
  • Patented Magic Arch™ support strip holds mask away from the nose and mouth to create a larger breathing chamber
  • Two styles to choose from:
    • Soft knit earloops
    • Anti-fog ear loops for use in high humidity environments or with protective eyewear
  • ASTM F2100 Compliant Level 1 mask
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 1
Color White
Fastening Type Earloops
Material Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Construction 3-ply
Disposable Single-use
Filter Efficiency ≥95% (PFE), ≥95% (BFE)
Includes Anti-fog earloops
Model WH 6155
Packaging Quantity 500/Cs.
Pore Size 0.1 μm
Resistance Fluid
Series CoolOne™