Learn More
AlphaProTech Critical Cover™ Coverall™ Face Shield
Catalog No. 17100906 Shop All AlphaProTech Products
Change view
Click to view available options
Material:
Polyester Film
Catalog No. Material
17-100-906 Polyester Film
1 options
Catalog No. 17-100-906 Supplier AlphaProTech Supplier No. 1807
Add to Cart
Add to Cart
The Coverall™ full face shield is lightweight, comfortable and protects against non-hazardous liquid splash and light particles.
  • Wrap-around design for clear peripheral vision
  • Protects against non-hazardous liquid splash and light particles in labs or controlled environments
  • Not ANSI Z87 rated for eye protection
  • Lightweight, comfortable fit for extended wear
  • Fits easily over prescription glasses or protective eyewear
  • Double-sided anti-fog coating minimizes fogging in high humidity
  • Low-distortion, 100% optically clear shield
  • Soft polyurethane foam headband for secure positioning
Specifications
Lens Coating Anti-fog
Lens Tint Clear
Product Type Full Coverage Faceshield
Material Polyester Film
Disposable Single-use
Lens Material None
Frame Material Rubber, Polyester
UV Protection None
Autoclavable Not Autoclavable