Learn More
AlphaProTech Critical Cover™ Coverall Face Shields
BIOPHARMA
Catalog No. 23309828 Shop All AlphaProTech Products
Change view
Click to view available options
Headband Material:
Adjustable Hook-and-Loop-Closure
Adjustable Hook-and-Loop-Closure Headband
Elastic Comfort Band
Model:
Full-face
Half-face
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Headband Material Model
23-309-828 Elastic Comfort Band Full-face
19-169-4021 Adjustable Hook-and-Loop-Closure Half-face
19-125-073 Elastic Comfort Band Half-face
23-309-829 Adjustable Hook-and-Loop-Closure Headband Full-face
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-309-828 Supplier AlphaProTech Supplier No. 2807
Add to Cart
Add to Cart
This item is not returnable. View return policy
Good peripheral vision while acting as a shield against dangerous splashes in the work environment
  • Lightweight and easily fits over glasses
  • Double-sided anti-fog coating
  • Low distorting and 100% optically clear
  • Soft polyurethane foam headband fits snugly against the forehead
  • Full-face style (Cat. Nos. 23-309-829 and 23-309-828) provides complete coverage including the sides of the face
Specifications
Lens Coating Anti-fog
Thickness (English) 7 mil shield
Description Fits easily over glasses
Lens Tint Clear
Headband Material Elastic Comfort Band
No. per Case 100/Cs.
Model Full-face