Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech Critical Cover™ GenPro™ Gowns
BIOPHARMA

Catalog No. 19166117 Shop All AlphaProTech Products
Change view
Click to view available options
Size:
Universal
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-166-117 X-Large
19-166-116 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-166-117 Supplier AlphaProTech Supplier No. GN135254
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Polypropylene gown with banded collar and waist ties provides a limited barrier to particulates and contamination in non-hazardous environments.

  • Comfortable
  • Lightweight
  • Moderate barrier protection
  • Exceptional breathability
  • Performs well in dry, non-hazardous environments
  • Fabric provides a limited barrier to particulates and contamination

Specifications

For Use With (Application) Clinical, Medical, Healthcare
Packaging Type Bulk
Product Type Isolation Gown
Style Full Coverage
Width (English) 30 in.
Length (English) 47 in.
Width (Metric) 762 mm
Gender Unisex
Length (Metric) 1200 mm
No. of Pockets None
Size X-Large
Material Polypropylene
Color Yellow
Closure Type Waist Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Latex Free Yes
Model GN-13525-4
Seam Type Serged
Sleeve Style Long Sleeve
Certifications/Compliance 21 CFR Part 820
Autoclavable Not Autoclavable
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.