Learn More
AlphaProTech Critical Cover™ GenPro™ Gown, Banded Ties at Waist and Neck
Shop All AlphaProTech ProductsDescription
- Comfortable
- Lightweight
- Banded ties are attached to the waist and neck for easy closure
- Moderate barrier protection
- Exceptional breathability
- Performs well in dry, non-hazardous environments
- Fabric provides a limited barrier to particulates and contamination
Specifications
Specifications
| Style | Full Coverage |
| Length (English) | 50 in. |
| Product Type | General Purpose Gown |
| Width (Metric) | 762 mm |
| Width (English) | 30 in. |
| Length (Metric) | 1270 mm |
| Size | X-Large |
| Gender | Unisex |
| No. of Pockets | None |
| Material | Polypropylene |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.