Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech Critical Cover™ GenPro™ Lab Coats
BIOPHARMA

Catalog No. 191304240 Shop All AlphaProTech Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Cuff Style:
Elastic
Length (English):
36.75 in.
39.5 in.
40 in.
42.75 in.
43.5 in.
44.5 in.
46.5 in.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Cuff Style Length (English)
19-130-4240 Large Elastic 40 in.
19-130-4241 X-Large Elastic 42.75 in.
19-130-4238 Small Elastic 36.75 in.
19-130-4243 3X-Large Elastic 44.5 in.
19-130-4239 Medium Elastic 39.5 in.
19-130-4242 2X-Large Elastic 43.5 in.
19-130-4244 4X-Large Elastic 46.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-130-4240 Supplier AlphaProTech Supplier No. LC124213
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Soft polypropylene lab coats designed for non-hazardous environments.

  • Designed for non-hazardous, general purpose environments
  • Ideal for personnel as well as facility visitors
  • Offer comfort and breathability
  • Full length, no-pocket design helps you control contaminants from entering or leaving sensitive areas

Specifications

Product Type Controlled Environments Lab Coat
Size Large
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets None
Collar Type Tapered
Seam Type Serged
Length (English) 40 in.
Length (Metric) 101.6 cm
Length (Metric) Sleeve 82.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.