Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech Critical Cover™ GenPro™ Shoe Covers: Serged Seam
BIOPHARMA

Catalog No. 191304271 Shop All AlphaProTech Products
Change view
Click to view available options
Size:
Universal
X-Large
Thread Color:
Black
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Thread Color
19-130-4271 X-Large Black
19-140-200 Universal White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-130-4271 Supplier AlphaProTech Supplier No. SH71223B
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Non-conductive polypropylene shoe covers are for use in non-hazardous environments. Ideal for general purpose or for facility visitors.

  • Moderate barrier to fluids, particulates and contamination
  • Available in anti-skid
  • Serged at the seam
  • Blue

Specifications

Size X-Large
Material Polypropylene
Latex Free No
Color Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top
Sole Type Anti-skid
Seam Type Serged
Thread Color Black
For Use With (Application) Non-hazardous Environments, Facility Visitors
Packaging Quantity 150 Pairs/Cs.
Length (English) 17.5 in.
Length (Metric) 44.45 cm
Height (English) 7 in.
Height (Metric) 17.78 cm
Resistance Dry Particulates, Fluid
Product Type Controlled Environments Shoe Covers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.