Learn More
Electron Microscopy Sciences Critical Point Dryer, Chamber Dimensions 63.5 mm internal diameter x 82 mm in length
SDP
Catalog No. NC3450750
Encompass_Preferred
Where increased chamber volume is required, for either size or quantity of specimens to be dried, the large capacity model E3100 is available. The chamber dimensions are 63.5 mm internal diameter x 82 mm in length, giving approximately three times the volume of the E3000. The transfer boat will also accept three times the number of specimen baskets.
Catalog No. NC3450750 Supplier Electron Microscopy Sciences Supplier No. E3100
Add to Cart
Add to Cart
Request a Quote
Where increased chamber volume is required, for either size or quantity of specimens to be dried, the large capacity model E3100 is available. The chamber dimensions are 63.5 mm internal diameter x 82 mm in length, giving approximately three times the volume of the E3000. The transfer boat will also accept three times the number of specimen baskets.