Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AquaPhoenix Critter and Creature Features Deluxe Kit, Bee an Engineer (Version 2) Deluxe Kit Components
SCI_ED

Catalog No. S39665MKC Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S39665MKC 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S39665MKC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Replacement parts for Educational Kit

Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.

Specifications

Color Assorted
For Use With (Application) Education kit
Quantity 100 Pk.
Length (English) 12 in.
Product Type Pipe cleaner
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.