Learn More
Selleck Chemical LLC crizotinib
SDP
Catalog No. 5000001461
Encompass_Preferred
(S)-crizotinib the (S)-enantiomer of crizotinib is a potent MTH1 (NUDT1) inhibitor with IC50 of 72 nM in a cell-free assay
Catalog No. 50-000-01461 Supplier Selleck Chemical LLC Supplier No. S750520MG
(S)-crizotinib the (S)-enantiomer of crizotinib is a potent MTH1 (NUDT1) inhibitor with IC50 of 72 nM in a cell-free assay