Learn More
Medchemexpress LLC Crizotinib | 877399-52-5 | 99.97% | C21H22Cl2FN5O | 1 ML
SDP
Catalog No. 5000357782
Encompass_Preferred
Crizotinib is an orally bioavailable, ATP-competitive inhibitor of ALK, c-Met, and ROS1. It inhibits tyrosine phosphorylation in cell-based assays and demonstrates effective tumor growth inhibition. This product is for research use only.
  • Orally bioavailable ALK, c-Met, and ROS1 inhibitor
  • Inhibits tyrosine phosphorylation of NPM-ALK and c-Met
  • Demonstrates effective tumor growth inhibition
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In-solvent storage: -80°C for 1 year; -20°C for 6 months
Catalog No. 50-003-57782 Supplier Medchemexpress LLC Supplier No. HY5087810MM/1ML
Crizotinib is an orally bioavailable, ATP-competitive inhibitor of ALK, c-Met, and ROS1. It inhibits tyrosine phosphorylation in cell-based assays and demonstrates effective tumor growth inhibition. This product is for research use only.
  • Orally bioavailable ALK, c-Met, and ROS1 inhibitor
  • Inhibits tyrosine phosphorylation of NPM-ALK and c-Met
  • Demonstrates effective tumor growth inhibition
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In-solvent storage: -80°C for 1 year; -20°C for 6 months