Learn More
Medchemexpress LLC Crizotinib (hydrochloride) | 1415560-69-8 | 99.7% | C21H23Cl3FN5O | 1 ML
SDP
Catalog No. 5000357799
Encompass_Preferred
Crizotinib hydrochloride is an orally bioavailable, selective, and ATP-competitive dual ALK and c-Met inhibitor. It inhibits tyrosine phosphorylation of NPM-ALK and c-Met in cell-based assays and is also a ROS proto-oncogene 1 (ROS1) inhibitor, demonstrating effective tumor growth inhibition.
  • Orally bioavailable
  • Selective ALK and c-Met inhibitor
  • Inhibits tyrosine phosphorylation of NPM-ALK and c-Met
  • Functions as a ROS proto-oncogene 1 (ROS1) inhibitor
  • Demonstrates effective tumor growth inhibition
Catalog No. 50-003-57799 Supplier Medchemexpress LLC Supplier No. HY50878A10MM/1ML
Crizotinib hydrochloride is an orally bioavailable, selective, and ATP-competitive dual ALK and c-Met inhibitor. It inhibits tyrosine phosphorylation of NPM-ALK and c-Met in cell-based assays and is also a ROS proto-oncogene 1 (ROS1) inhibitor, demonstrating effective tumor growth inhibition.
  • Orally bioavailable
  • Selective ALK and c-Met inhibitor
  • Inhibits tyrosine phosphorylation of NPM-ALK and c-Met
  • Functions as a ROS proto-oncogene 1 (ROS1) inhibitor
  • Demonstrates effective tumor growth inhibition