Learn More
Selleck Chemical LLC Crizotinib (PF-02341066) 5mg 877399-52-5
SDP
Catalog No. 508270
Encompass_Preferred
Crizotinib (PF-02341066) is a potent inhibitor of c-Met and ALK with IC50 of 11 nM and 24 nM in cell-based assays, respectively. It is also a potent ROS1 inhibitor with Ki value less than 0.025 nM. Crizotinib induces autophagy through inhibition of the STAT3 pathway in multiple lung cancer cell lines. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-827-0 Supplier Selleck Chemical LLC Supplier No. S10685MG
Crizotinib (PF-02341066) is a potent inhibitor of c-Met and ALK with IC50 of 11 nM and 24 nM in cell-based assays, respectively. It is also a potent ROS1 inhibitor with Ki value less than 0.025 nM. Crizotinib induces autophagy through inhibition of the STAT3 pathway in multiple lung cancer cell lines. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.