Learn More
TARGETMOL CHEMICALS INC CRK12-IN-2 5MG
SDP
Catalog No. 502579409
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CRK12-IN-2 is a potent inhibitor of CRK12.CRK12-IN-2 inhibits the parasite with EC50 values of 3.2 and 0.08 nM for Trypanosoma congolense and Trypanosoma vivax respectively.CRK12-IN-2 can be used for the prevention and treatment of trypanosomiasis in animals. purity: 99%
Catalog No. 50-257-9409 Supplier TARGETMOL CHEMICALS INC Supplier No. T637275MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CRK12-IN-2 is a potent inhibitor of CRK12.CRK12-IN-2 inhibits the parasite with EC50 values of 3.2 and 0.08 nM for Trypanosoma congolense and Trypanosoma vivax respectively.CRK12-IN-2 can be used for the prevention and treatment of trypanosomiasis in animals. purity: 99%