Learn More
Aviva Systems Biology Corp CRLF1 Antibody - middle region (ARP36673_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36673_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRLF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CRLF1. Alternate names: CISS, CISS1, CLF, CLF-1, NR6, zcytor5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-56244