Learn More
Sino Biological Human CRLF2 Protein (ECD, Fc Tag) 50ug
Supplier:  Sino Biological 29749H02H50
Encompass_Preferred
A DNA sequence encoding the human CRLF2 (NP_071431.2) (Met1-Lys231) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-5621