Learn More
Aviva Systems Biology Corp CRLS1 Antibody - middle region (ARP49444_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49444_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRLS1. It was validated on Western Blot. CRLS1,cardiolipin synthase 1,CLS; CLS1; GCD10; C20orf155; dJ967N21.6; CRLS1,cardiolipin synthase,cardiolipin synthaseThe immunogen is a synthetic peptide directed towards the middle region of human CRLS1. Alternate names: C20orf155, CLS1, GCD10, dJ967N21.6, CLSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-422