Learn More
Aviva Systems Biology Corp CRMP1 Antibody - N-terminal region (ARP41933_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41933_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRMP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CRMP1. Alternate names: DPYSL1, DRP-1, DRP1, CRMP-1, ULIP-3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58054