Learn More
ECM Bioscience CRMP2 (Ser-522), phospho-specific Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience CP2191
Encompass_Preferred
This antibody was affinity purified using CRMP2 (Ser-522) peptide (without carrier). The antibody detects a 70 kDa* protein corresponding to the molecular mass of CRMP2 on SDS-PAGE immunoblots of human mouse brain, and this band is removed after lambda phosphatase treatment. In addition, this reactivity can be specifically blocked using phospho-CRMP2 (Ser-522) peptide (CX2195). The immunogen was Phospho-CRMP2 (Ser-522) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding Ser-522 in human CRMP2. This sequence is conserved in rat and mouse CRMP2, and has homology to the conserved site in CRMP1 and CRMP4, but is not conserved in CRMP3 and CRMP5. Accession #Q16555. Applications = WB, E, ICC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = DRP-2, Toad-64, CRMP-62
Catalog No. 50-172-0748