Learn More
Aviva Systems Biology Corp CRNN Antibody - middle region (ARP42460_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42460_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRNN. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CRNN. Alternate names: C1orf10, PDRC1, SEP53, DRC1ApplicationWB. Species reactivity: Dog, Horse, Human
Catalog No. 50-285-004