Learn More
Aviva Systems Biology Corp CROCCL2 Antibody - N-terminal region (ARP51734_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51734_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CROCCL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CROCCL2. Alternate names: KIAA0445, KIAA1922, dJ798A10.2, CROCCL2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53667