Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Crocetin dialdehyde | 502-70-5 | 296.40 | 1 MG
SDP

Catalog No. 5000416574
Encompass_Preferred

Crocetin dialdehyde is a non-volatile apocarotenoid obtained by the action of the plastidic enzyme CCD2 over the carotenoid zeaxanthin in saffron. It can be converted into Crocetin by aldehyde dehydrogenases (ALDHs) and has shown no effect on the liver in mice.

  • Purity: 99.21%
  • Appearance: Solid
  • Color: Orange to reddish brown
  • Initial source: Plants Iridaceae Crocus sativus Linn.
  • For research use only.
  • Shown no effect on the liver in mice.

Catalog No. 50-004-16574 Supplier Medchemexpress LLC Supplier No. HYN109911MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crocetin dialdehyde is a non-volatile apocarotenoid obtained by the action of the plastidic enzyme CCD2 over the carotenoid zeaxanthin in saffron. It can be converted into Crocetin by aldehyde dehydrogenases (ALDHs) and has shown no effect on the liver in mice.

  • Purity: 99.21%
  • Appearance: Solid
  • Color: Orange to reddish brown
  • Initial source: Plants Iridaceae Crocus sativus Linn.
  • For research use only.
  • Shown no effect on the liver in mice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.