Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Crocetin dialdehyde | 502-70-5 | 296.40 | 5 MG
SDP

Catalog No. 5000414765
Encompass_Preferred

Crocetin dialdehyde is a non-volatile apocarotenoid. It is derived from the action of the plastidic enzyme CCD2 on the carotenoid zeaxanthin, found in saffron. Crocetin dialdehyde can be converted into Crocetin (HY-N2072) by aldehyde dehydrogenases (ALDHs). Studies in mice have shown that Crocetin dialdehyde has no effect on the liver.

  • Non-volatile apocarotenoid.
  • Can be converted to Crocetin by ALDHs.
  • No observed effect on the liver in mice (at 300 mg/kg, p.o., for 1 week).
  • Purity: 99.21%.
  • Appearance: Solid, orange to reddish brown.

Catalog No. 50-004-14765 Supplier Medchemexpress LLC Supplier No. HYN109915MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crocetin dialdehyde is a non-volatile apocarotenoid. It is derived from the action of the plastidic enzyme CCD2 on the carotenoid zeaxanthin, found in saffron. Crocetin dialdehyde can be converted into Crocetin (HY-N2072) by aldehyde dehydrogenases (ALDHs). Studies in mice have shown that Crocetin dialdehyde has no effect on the liver.

  • Non-volatile apocarotenoid.
  • Can be converted to Crocetin by ALDHs.
  • No observed effect on the liver in mice (at 300 mg/kg, p.o., for 1 week).
  • Purity: 99.21%.
  • Appearance: Solid, orange to reddish brown.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.