Learn More
Medchemexpress LLC Crocetin disodium | 112108-01-7 | 99.3% | 372.37 | 50 MG
SDP
Catalog No. 5000394675
Encompass_Preferred
Crocetin disodium, extracted from saffron (Crocus sativus L.), acts as an NMDA receptor antagonist with high affinity. It is a saffron metabolite originating from crocin apocarotenoids and has shown strong NMDA receptor affinity, believed to be responsible for the CNS activity of saffron.
  • Extracted from saffron (Crocus sativus L.)
  • Acts as an NMDA receptor antagonist with high affinity
  • Saffron metabolite originating from crocin apocarotenoids
  • For research use only
Catalog No. 50-003-94675 Supplier Medchemexpress LLC Supplier No. HY1650250MG
Crocetin disodium, extracted from saffron (Crocus sativus L.), acts as an NMDA receptor antagonist with high affinity. It is a saffron metabolite originating from crocin apocarotenoids and has shown strong NMDA receptor affinity, believed to be responsible for the CNS activity of saffron.
  • Extracted from saffron (Crocus sativus L.)
  • Acts as an NMDA receptor antagonist with high affinity
  • Saffron metabolite originating from crocin apocarotenoids
  • For research use only