Learn More
Medchemexpress LLC Crocetin (disodium) | 591230-99-8 | 99.3% | 372.37 | 1 ML
SDP
Catalog No. 5000394447
Encompass_Preferred
Crocetin disodium, extracted from saffron (Crocus sativus L.), acts as an NMDA receptor antagonist with high affinity. It is a saffron metabolite originating from the crocin apocarotenoids and has been shown to exert strong NMDA receptor affinity, which is thought to be responsible for the CNS activity of saffron. This solid, orange to reddish brown compound is for research use only.
  • Acts as an NMDA receptor antagonist with high affinity.
  • Derived from saffron (Crocus sativus L.) and is a saffron metabolite.
  • Responsible for CNS activity of saffron.
  • Soluble in H2O: ≥ 4.56 mg/mL (12.25 mM).
  • Solid appearance, orange to reddish brown color.
Catalog No. 50-003-94447 Supplier Medchemexpress LLC Supplier No. HY1650210MM/1ML
Crocetin disodium, extracted from saffron (Crocus sativus L.), acts as an NMDA receptor antagonist with high affinity. It is a saffron metabolite originating from the crocin apocarotenoids and has been shown to exert strong NMDA receptor affinity, which is thought to be responsible for the CNS activity of saffron. This solid, orange to reddish brown compound is for research use only.
  • Acts as an NMDA receptor antagonist with high affinity.
  • Derived from saffron (Crocus sativus L.) and is a saffron metabolite.
  • Responsible for CNS activity of saffron.
  • Soluble in H2O: ≥ 4.56 mg/mL (12.25 mM).
  • Solid appearance, orange to reddish brown color.