Learn More
Medchemexpress LLC Crocetin disodium | 591230-99-8 | 99.3% | 372.37 | 100 MG
SDP
Catalog No. 5000394211
Encompass_Preferred
Crocetin (Transcrocetin) disodium is extracted from saffron (Crocus sativus L.) and acts as an NMDA receptor antagonist with high affinity. It is for research use only and not sold to patients.
  • Extracted from saffron (Crocus sativus L.).
  • Acts as an NMDA receptor antagonist.
  • Exhibits high affinity.
Catalog No. 50-003-94211 Supplier Medchemexpress LLC Supplier No. HY16502100MG
Crocetin (Transcrocetin) disodium is extracted from saffron (Crocus sativus L.) and acts as an NMDA receptor antagonist with high affinity. It is for research use only and not sold to patients.
  • Extracted from saffron (Crocus sativus L.).
  • Acts as an NMDA receptor antagonist.
  • Exhibits high affinity.