Learn More
Medchemexpress LLC Crocetin disodium | 591230-99-8 | 99.3% | 372.37 | 5 MG
SDP
Catalog No. 5000393985
Encompass_Preferred
Crocetin disodium, extracted from saffron (Crocus sativus L.), acts as an NMDA receptor antagonist with high affinity.
  • Acts as an NMDA receptor antagonist.
  • Exerts strong NMDA receptor affinity.
  • Responsible for the central nervous system (CNS) activity of saffron.
Catalog No. 50-003-93985 Supplier Medchemexpress LLC Supplier No. HY165025MG
Crocetin disodium, extracted from saffron (Crocus sativus L.), acts as an NMDA receptor antagonist with high affinity.
  • Acts as an NMDA receptor antagonist.
  • Exerts strong NMDA receptor affinity.
  • Responsible for the central nervous system (CNS) activity of saffron.