Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Crocetin disodium | 591230-99-8 | MFCD22572782 | 99.3% | 372.37 g/mol | C20H22Na2O4 | 10 MG
SDP

Catalog No. 5000289266
Encompass_Preferred

Crocetin disodium is the disodium salt of crocetin, a carotenoid extracted from saffron (Crocus sativus L.). It acts as an ionotropic glutamate (iGluR) antagonist with high affinity for NMDA receptors and is supplied for in vitro research use.

  • Acts as an NMDA (iGluR) receptor antagonist with high affinity.
  • High purity, reported as 99.3%.
  • Chemical formula C20H22Na2O4; molecular weight 372.37 g/mol.
  • Orange to reddish-brown solid; also available as an aqueous solution (10 mM, 1 mL).
  • Soluble in water (≥ 4.56 mg/mL, ~12.25 mM); insoluble or slightly soluble in DMSO (< 1 mg/mL).
  • Intended for in vitro research applications.

Catalog No. 50-002-89266 Supplier Medchemexpress LLC Supplier No. HY1650210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crocetin disodium is the disodium salt of crocetin, a carotenoid extracted from saffron (Crocus sativus L.). It acts as an ionotropic glutamate (iGluR) antagonist with high affinity for NMDA receptors and is supplied for in vitro research use.

  • Acts as an NMDA (iGluR) receptor antagonist with high affinity.
  • High purity, reported as 99.3%.
  • Chemical formula C20H22Na2O4; molecular weight 372.37 g/mol.
  • Orange to reddish-brown solid; also available as an aqueous solution (10 mM, 1 mL).
  • Soluble in water (≥ 4.56 mg/mL, ~12.25 mM); insoluble or slightly soluble in DMSO (< 1 mg/mL).
  • Intended for in vitro research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.