Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Crocetin monomethyl ester | 25368-09-6 | 1 MG
SDP

Catalog No. 5000417902
Encompass_Preferred

Crocetin monomethyl ester, isolated from Crocus sativus, possesses anti-inflammatory, neuroprotective, and antioxidant activities. It promotes the clearance of amyloid-β by inducing autophagy via the STK11/LKB1-mediated AMPK pathway.

  • Isolated from Crocus sativus.
  • Possesses anti-inflammatory, neuroprotective, and antioxidant activities.
  • Promotes the clearance of amyloid-β by inducing autophagy through the STK11/LKB1-mediated AMPK pathway.
  • For research use only.
  • Purity of 98.0% to 98.44%.
  • Appears as a solid, brown to reddish-brown.
  • Soluble in DMSO (31.25 mg/mL).
  • Store at -20°C, sealed, away from moisture and light. In solvent, store at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-004-17902 Supplier Medchemexpress LLC Supplier No. HYN69041MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crocetin monomethyl ester, isolated from Crocus sativus, possesses anti-inflammatory, neuroprotective, and antioxidant activities. It promotes the clearance of amyloid-β by inducing autophagy via the STK11/LKB1-mediated AMPK pathway.

  • Isolated from Crocus sativus.
  • Possesses anti-inflammatory, neuroprotective, and antioxidant activities.
  • Promotes the clearance of amyloid-β by inducing autophagy through the STK11/LKB1-mediated AMPK pathway.
  • For research use only.
  • Purity of 98.0% to 98.44%.
  • Appears as a solid, brown to reddish-brown.
  • Soluble in DMSO (31.25 mg/mL).
  • Store at -20°C, sealed, away from moisture and light. In solvent, store at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.