Learn More
Medchemexpress LLC Crocetin monomethyl ester | 25368-09-6 | 1 ML
SDP
Catalog No. 5000418348
Encompass_Preferred
Crocetin monomethyl ester, isolated from *Crocus sativus*, possesses anti-inflammatory, neuroprotective, and antioxidant activity. It promotes the clearance of amyloid-β by inducing autophagy via the STK11/LKB1-mediated AMPK pathway.
  • Anti-inflammatory activity
  • Neuroprotective activity
  • Antioxidant activity
  • Promotes clearance of amyloid-β
  • Induces autophagy via STK11/LKB1-mediated AMPK pathway
  • Isolated from *Crocus sativus*
  • Minimum 98% purity
Catalog No. 50-004-18348 Supplier Medchemexpress LLC Supplier No. HYN690410MM/1ML
Crocetin monomethyl ester, isolated from *Crocus sativus*, possesses anti-inflammatory, neuroprotective, and antioxidant activity. It promotes the clearance of amyloid-β by inducing autophagy via the STK11/LKB1-mediated AMPK pathway.
  • Anti-inflammatory activity
  • Neuroprotective activity
  • Antioxidant activity
  • Promotes clearance of amyloid-β
  • Induces autophagy via STK11/LKB1-mediated AMPK pathway
  • Isolated from *Crocus sativus*
  • Minimum 98% purity