Learn More
Medchemexpress LLC Crocin | 42553-65-1 | 976.96 | 5 MG
SDP
Catalog No. 5000416795
Encompass_Preferred
Crocin is an orally active natural product isolated from the stigma of Crocus sativus. It inhibits tumor cell proliferation and promotes apoptosis through the JAK pathway, while also exhibiting anti-inflammatory, antioxidant, and antitumor activities.
  • Isolated from the stigma of Crocus sativus
  • Inhibits tumor cell proliferation and promotes apoptosis
  • Exhibits anti-inflammatory, antioxidant, and antitumor activities
  • Weakens colon cancer cell vitality in a concentration-dependent manner
  • Facilitates apoptosis of HCT-116 cells
  • Decreases levels of key signaling proteins (p-JAK2, p-STAT3, p-ERK)
  • Shows antioxidant, anti-inflammatory, and antifibrotic effects in rat models
  • Reduces expression of TLR4, IL-10, TNF-α, and TGF-β1
  • Purity of 99.30%
  • Appears as a solid, brown to reddish brown substance
Catalog No. 50-004-16795 Supplier Medchemexpress LLC Supplier No. HYN06975MG
Add to Cart
Add to Cart
Crocin is an orally active natural product isolated from the stigma of Crocus sativus. It inhibits tumor cell proliferation and promotes apoptosis through the JAK pathway, while also exhibiting anti-inflammatory, antioxidant, and antitumor activities.
  • Isolated from the stigma of Crocus sativus
  • Inhibits tumor cell proliferation and promotes apoptosis
  • Exhibits anti-inflammatory, antioxidant, and antitumor activities
  • Weakens colon cancer cell vitality in a concentration-dependent manner
  • Facilitates apoptosis of HCT-116 cells
  • Decreases levels of key signaling proteins (p-JAK2, p-STAT3, p-ERK)
  • Shows antioxidant, anti-inflammatory, and antifibrotic effects in rat models
  • Reduces expression of TLR4, IL-10, TNF-α, and TGF-β1
  • Purity of 99.30%
  • Appears as a solid, brown to reddish brown substance