Learn More
Medchemexpress LLC Crocin (Crocin I) | 42553-65-1 | MFCD00017495 | 98.5% | 976.96 | C44H64O24 | 10 MG
SDP
Catalog No. 5000295731
Encompass_Preferred
Crocin is a natural carotenoid glycoside isolated from the stigma of Crocus sativus. It is used as a research reagent with reported anti-inflammatory, antioxidant, and antitumor activities and has been reported to inhibit tumor cell proliferation and induce apoptosis via the JAK signaling pathway.
  • Natural carotenoid isolated from Crocus sativus.
  • Orally active compound used in biochemical and cell biology research.
  • Reported activities include anti-inflammatory, antioxidant, and antitumor effects.
  • Inhibits tumor cell proliferation and promotes apoptosis via the JAK pathway.
  • Chemical identifiers: CAS 42553-65-1; formula C44H64O24; molecular weight 976.96.
  • Available in small research pack sizes suitable for laboratory use.
Catalog No. 50-002-95731 Supplier Medchemexpress LLC Supplier No. HYN069710MG
Add to Cart
Add to Cart
Crocin is a natural carotenoid glycoside isolated from the stigma of Crocus sativus. It is used as a research reagent with reported anti-inflammatory, antioxidant, and antitumor activities and has been reported to inhibit tumor cell proliferation and induce apoptosis via the JAK signaling pathway.
  • Natural carotenoid isolated from Crocus sativus.
  • Orally active compound used in biochemical and cell biology research.
  • Reported activities include anti-inflammatory, antioxidant, and antitumor effects.
  • Inhibits tumor cell proliferation and promotes apoptosis via the JAK pathway.
  • Chemical identifiers: CAS 42553-65-1; formula C44H64O24; molecular weight 976.96.
  • Available in small research pack sizes suitable for laboratory use.