Learn More
TARGETMOL CHEMICALS INC CROCIN II 10MG
SDP
Catalog No. 502580125
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Crocin II (Crocetin gentiobiosylglucosyl ester) has anti-inflammatory and anti-apoptotic effects against gastric ischemia-reperfusion injury in rats the effect of crocin is mainly mediated by reducing the protein expression of iNOS and caspase-3. Crocin II can ameliorate CCl4-induced liver injury via inhibition of inflammatory cytokines caspase3 and oxidative stress along with modulation of liver metabolizing enzymes favoring elimination of CCl4 toxic metabolite. Crocin II has antihyperlipidemic effect may be due to the inhibition of pancreatic lipase and crocin and its metabolite Crocin II can improve hyperlipidemia. Crocin has antidepressant effects by increasing CREB BDNF and VGF levels in hippocampus. purity: 99%
Catalog No. 50-258-0125 Supplier TARGETMOL CHEMICALS INC Supplier No. T377910MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Crocin II (Crocetin gentiobiosylglucosyl ester) has anti-inflammatory and anti-apoptotic effects against gastric ischemia-reperfusion injury in rats the effect of crocin is mainly mediated by reducing the protein expression of iNOS and caspase-3. Crocin II can ameliorate CCl4-induced liver injury via inhibition of inflammatory cytokines caspase3 and oxidative stress along with modulation of liver metabolizing enzymes favoring elimination of CCl4 toxic metabolite. Crocin II has antihyperlipidemic effect may be due to the inhibition of pancreatic lipase and crocin and its metabolite Crocin II can improve hyperlipidemia. Crocin has antidepressant effects by increasing CREB BDNF and VGF levels in hippocampus. purity: 99%