Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Crocin II | 55750-84-0 | MFCD11111454 | 98.0% | 10 MG
SDP

Catalog No. 5000295661
Encompass_Preferred

Crocin II is a natural glycoside isolated from Gardenia jasminoides, supplied as a solid research reagent and analytical standard with reported antioxidant, anticancer, and antidepressant activities. The product is characterized by a molecular weight of 814.82 g/mol, CAS number 55750-84-0, and a recorded purity around 98%. Recommended storage is -20°C and protect from light; solubility in DMSO is approximately 25 mg/mL.

  • Natural glycoside isolated from Gardenia jasminoides
  • High purity suitable for analytical applications
  • Molecular weight 814.82 g/mol and CAS 55750-84-0
  • Soluble in DMSO (≈25 mg/mL) for in vitro use
  • Storage recommended at -20°C and protect from light

Catalog No. 50-002-95661 Supplier Medchemexpress LLC Supplier No. HYN069810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crocin II is a natural glycoside isolated from Gardenia jasminoides, supplied as a solid research reagent and analytical standard with reported antioxidant, anticancer, and antidepressant activities. The product is characterized by a molecular weight of 814.82 g/mol, CAS number 55750-84-0, and a recorded purity around 98%. Recommended storage is -20°C and protect from light; solubility in DMSO is approximately 25 mg/mL.

  • Natural glycoside isolated from Gardenia jasminoides
  • High purity suitable for analytical applications
  • Molecular weight 814.82 g/mol and CAS 55750-84-0
  • Soluble in DMSO (≈25 mg/mL) for in vitro use
  • Storage recommended at -20°C and protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.