Learn More
Cayman Chemical Crocin/Gardenia Extract, CAS: 42553-65-1, 5 g
SDP
Catalog No. 502935381
Encompass_Preferred
A Gardenia extract that contains crocin I, II, and III, and crocetin, among other compounds.
Catalog No. 50-293-5381 Supplier Cayman Chemical Supplier No. 263375 G
A Gardenia extract that contains crocin I, II, and III, and crocetin, among other compounds.