Learn More
TARGETMOL CHEMICALS INC CROCONAZOLE 10MG
SDP
Catalog No. 502578944
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Croconazole exhibited dose-dependent inhibitory activity on the 5-lipoxygenase (5-LOX) of neutrophils. Croconazole is an antimycotic agent. The IC50s for the synthesis of leukotriene B4 (LTB4) and 5-hydroxyeicosatetraenoic acid (5-HETE) is 7.8 +/- 1.7 andpurity: 99%
Catalog No. 50-257-8944 Supplier TARGETMOL CHEMICALS INC Supplier No. T987810MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Croconazole exhibited dose-dependent inhibitory activity on the 5-lipoxygenase (5-LOX) of neutrophils. Croconazole is an antimycotic agent. The IC50s for the synthesis of leukotriene B4 (LTB4) and 5-hydroxyeicosatetraenoic acid (5-HETE) is 7.8 +/- 1.7 andpurity: 99%