Learn More
Medchemexpress LLC Croconic acid disod 50mg
SDP
Catalog No. 5000204904
Encompass_Preferred
Croconic acid (disodium) (Nacr) can promote cell growth and proliferation by enhancing the expression of genes associated with lysine crotonylation (Kcr) modification Croconic acid holds potential for improving somatic cell nuclear transfer (SCNT) efficiency and optimizing cell culture conditions research[1]
Catalog No. 50-002-04904 Supplier Medchemexpress LLC Supplier No. HYW105310A50MG
Add to Cart
Add to Cart
Croconic acid (disodium) (Nacr) can promote cell growth and proliferation by enhancing the expression of genes associated with lysine crotonylation (Kcr) modification Croconic acid holds potential for improving somatic cell nuclear transfer (SCNT) efficiency and optimizing cell culture conditions research[1]