Learn More
Trajan Scientific CROM LNR 2.3MM 5PK
SDP
Catalog No. 502530185
Encompass_Preferred
SGE GC Inlet FAST FocusLiner, Diameter Inner: 0.23 cm, Diameter Outer: 0.63 cm, Length: 7.85 cm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, Packing Material: Quartz Wool, Split/Splitless Fast Focus Liner, Quantity: 5/Pk.
Catalog No. 50-253-0185 Supplier Trajan Scientific Supplier No. 092005
SGE GC Inlet FAST FocusLiner, Diameter Inner: 0.23 cm, Diameter Outer: 0.63 cm, Length: 7.85 cm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, Packing Material: Quartz Wool, Split/Splitless Fast Focus Liner, Quantity: 5/Pk.