Learn More
Trajan Scientific CROM LNR TAPFCS 25PK
SDP
Catalog No. 502530184
Encompass_Preferred
Trajan Split / Splitless Tapered FocusLiner for Agilent, Quantity: 25/Pk., Diameter Inner: 4 mm, Diameter Outer: 6.3 mm, Length: 78.5 mm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, GC Inlet Liner, Split/Splitless Tapered FocusLiner
Catalog No. 50-253-0184 Supplier Trajan Scientific Supplier No. 092011
Trajan Split / Splitless Tapered FocusLiner for Agilent, Quantity: 25/Pk., Diameter Inner: 4 mm, Diameter Outer: 6.3 mm, Length: 78.5 mm, For Use With: Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177, GC Inlet Liner, Split/Splitless Tapered FocusLiner