Learn More
Cayman Chemical (±)-Cromakalim, Purity: ≥98%, CAS: 94470-67-4, 50 mg
SDP
Catalog No. 502972868
Encompass_Preferred
An activator of the ATP-sensitive K+ channels that displays anti-ischemic (EC25 = 8.9 μM) and cardioprotective (IC50 = 0.03 μM) effects in rat hearts
Catalog No. 50-297-2868 Supplier Cayman Chemical Supplier No. 1460150 MG
An activator of the ATP-sensitive K+ channels that displays anti-ischemic (EC25 = 8.9 μM) and cardioprotective (IC50 = 0.03 μM) effects in rat hearts