Learn More
Abcam Cromakalim, KIR6. x (KATP) channel activator, 50MG
Catalog No. 502707559
Encompass_Preferred
MW 286.33 g/mol, Purity >99%. KIR6.x (KATP) channel activator. Orally active, potent smooth muscle relaxant.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7559 Supplier Abcam Supplier No. AB12048650MG
MW 286.33 g/mol, Purity >99%. KIR6.x (KATP) channel activator. Orally active, potent smooth muscle relaxant.
The product is subject to the following: Abcam Restricted Use Statement