Learn More
Medchemexpress LLC Cromolyn disodium | 15826-37-6 | 100.0% | 1 G
SDP
Catalog No. 5000406662
Encompass_Preferred
Cromolyn disodium (Cromoglycate disodium) is an orally active GSK-3β inhibitor and a mast cell stabilizer. It inhibits the release of mediators from mast cells, regulates reflex bronchoconstriction, and reduces non-specific bronchial hyperreactivity, making it useful in the research of bronchial asthma. This compound also demonstrates anti-inflammatory, anti-allergic, anti-histamine, anti-cancer, and neuroprotective effects.
  • Orally active GSK-3β inhibitor
  • Mast cell stabilizer
  • Inhibits release of mediators from mast cells
  • Regulates reflex bronchoconstriction
  • Reduces non-specific bronchial hyperreactivity
  • Exhibits anti-inflammatory properties
  • Exhibits anti-allergic properties
  • Exhibits anti-histamine properties
  • Demonstrates anti-cancer effects
  • Provides neuroprotective effects
Catalog No. 50-004-06662 Supplier Medchemexpress LLC Supplier No. HYB0320A1G
Cromolyn disodium (Cromoglycate disodium) is an orally active GSK-3β inhibitor and a mast cell stabilizer. It inhibits the release of mediators from mast cells, regulates reflex bronchoconstriction, and reduces non-specific bronchial hyperreactivity, making it useful in the research of bronchial asthma. This compound also demonstrates anti-inflammatory, anti-allergic, anti-histamine, anti-cancer, and neuroprotective effects.
  • Orally active GSK-3β inhibitor
  • Mast cell stabilizer
  • Inhibits release of mediators from mast cells
  • Regulates reflex bronchoconstriction
  • Reduces non-specific bronchial hyperreactivity
  • Exhibits anti-inflammatory properties
  • Exhibits anti-allergic properties
  • Exhibits anti-histamine properties
  • Demonstrates anti-cancer effects
  • Provides neuroprotective effects